4-Iodo-2-methoxypyridine 98%

BLDPBD38126-250MGEA 17 EUR
BLDPBD38126-250MG BLDPBD38126-1G BLDPBD38126-25G BLDPBD38126-0.025G BLDPBD38126-5G
4-Iodo-2-methoxypyridine 98%
4-Iodo-2-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₆INO
MW: 235,0224 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD08277272
CAS-nummer: 98197-72-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR