1-(2-Bromophenyl)piperidine 97%

BLDPBD40612-1GEA 15 EUR
BLDPBD40612-1G BLDPBD40612-0.025G BLDPBD40612-5G BLDPBD40612-25G
1-(2-Bromophenyl)piperidine 97%
1-(2-Bromophenyl)piperidine

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₁H₁₄BrN
MW: 240,1396 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04112586
CAS-nummer: 156808-79-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR