4-Chloro-6-ethoxy-2-pyrimidinamine 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD40655-250MGEA 48 EUR
BLDPBD40655-250MG BLDPBD40655-5G BLDPBD40655-1G
4-Chloro-6-ethoxy-2-pyrimidinamine 95%
4-Chloro-6-ethoxy-2-pyrimidinamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₆H₈ClN₃O
MW: 173,6000 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00457060
CAS-nummer: 89784-02-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR