3-Bromo-1H-pyrazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD40744-1GEA 9 EUR
BLDPBD40744-1G BLDPBD40744-250MG BLDPBD40744-10G BLDPBD40744-100G BLDPBD40744-25G BLDPBD40744-5G
3-Bromo-1H-pyrazole 98%
3-Bromo-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formule: C3H3BrN2
MW: 146,97 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD01646049
CAS-nummer: 14521-80-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR