6-Chloropyridazine-3-carbonitrile 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD40752-100MGEA 26 EUR
BLDPBD40752-100MG BLDPBD40752-5G BLDPBD40752-10G BLDPBD40752-250MG BLDPBD40752-1G
6-Chloropyridazine-3-carbonitrile 97%
6-Chloropyridazine-3-carbonitrile

Storage condition: Inert atmosphere, 2-8°C

Formule: C₅H₂ClN₃
MW: 139,5400 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD09835352
CAS-nummer: 35857-89-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR