2,3,6-Trifluorobenzoic acid 98%

BLDPBD4085-100GEA 297 EUR
BLDPBD4085-100G BLDPBD4085-1G BLDPBD4085-25G BLDPBD4085-5G BLDPBD4085-10G BLDPBD4085-250MG
2,3,6-Trifluorobenzoic acid 98%
2,3,6-Trifluorobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₃F₃O₂
MW: 176,09 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00002409
CAS-nummer: 2358-29-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR