1-Methylimidazool 98%

Synoniemen: NMI

BLDPBD40860-0.025GEA 0 EUR
BLDPBD40860-0.025G BLDPBD40860-25G
1-Methylimidazool 98%
1-Methylimidazool

Storage condition: Inert atmosphere, Room Temperature

Formule: C₄H₆N₂
MW: 82,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005292
CAS-nummer: 616-47-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR