2-Aminoacetonitrile hydrochloride 98%

BLDPBD40871-25GEA 7 EUR
BLDPBD40871-25G BLDPBD40871-10G
2-Aminoacetonitrile hydrochloride 98%
2-Aminoacetonitrile hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₂H₅ClN₂
MW: 92,53 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00012850
CAS-nummer: 6011-14-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR