6-Chloro-m-toluic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD40950-10GEA 57 EUR
BLDPBD40950-10G BLDPBD40950-1G BLDPBD40950-25G BLDPBD40950-5G BLDPBD40950-100G
6-Chloro-m-toluic acid 97%
6-Chloro-m-toluic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₈H₇ClO₂
MW: 170,6 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00045798
CAS-nummer: 6342-60-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR