2-Bromo-6-chlorobenzoic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD4124-500GEA 733 EUR
BLDPBD4124-500G BLDPBD4124-25G BLDPBD4124-10G BLDPBD4124-1G BLDPBD4124-100G BLDPBD4124-0.025G BLDPBD4124-5G
2-Bromo-6-chlorobenzoic acid 98%
2-Bromo-6-chlorobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₄BrClO₂
MW: 235,46 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00672929
CAS-nummer: 93224-85-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR