4,5-Diamino-6-hydroxypyrimidine 95%

BLDPBD41361-250MGEA 7 EUR
BLDPBD41361-250MG BLDPBD41361-1G BLDPBD41361-10G BLDPBD41361-5G BLDPBD41361-25G
4,5-Diamino-6-hydroxypyrimidine 95%
4,5-Diamino-6-hydroxypyrimidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₄H₆N₄O
MW: 126,1200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00006114
CAS-nummer: 1672-50-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR