2,3-Difluorotoluene 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD41645-10GEA 29 EUR
BLDPBD41645-10G BLDPBD41645-1G BLDPBD41645-25G BLDPBD41645-5G
2,3-Difluorotoluene 98%
2,3-Difluorotoluene

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₆F₂
MW: 128,12 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01631506
CAS-nummer: 3828-49-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR