3-Chloro-o-xylene 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD41850-1GEA 6 EUR
BLDPBD41850-1G BLDPBD41850-100G BLDPBD41850-5G BLDPBD41850-25G
3-Chloro-o-xylene 98%
3-Chloro-o-xylene

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₉Cl
MW: 140,61 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00060689
CAS-nummer: 608-23-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR