3-Fluoro-4-nitrobenzoic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD4248-1GEA 7 EUR
BLDPBD4248-1G BLDPBD4248-100G BLDPBD4248-25G BLDPBD4248-10G BLDPBD4248-5G BLDPBD4248-500G
3-Fluoro-4-nitrobenzoic acid 98%
3-Fluoro-4-nitrobenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₄FNO₄
MW: 185,11 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01862092
CAS-nummer: 403-21-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR