2-Hydroxydimetridazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD43612-1GEA 27 EUR
BLDPBD43612-1G BLDPBD43612-100G BLDPBD43612-250MG BLDPBD43612-5G BLDPBD43612-25G
2-Hydroxydimetridazole 98%
2-Hydroxydimetridazole

Storage condition: Sealed in dry, Room Temperature

Formule: C₅H₇N₃O₃
MW: 157,1274 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00159675
CAS-nummer: 936-05-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR