4-Iodo-2-nitrotoluene 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD4654-1GEA 6 EUR
BLDPBD4654-1G BLDPBD4654-500G BLDPBD4654-25G BLDPBD4654-100G BLDPBD4654-10G BLDPBD4654-5G
4-Iodo-2-nitrotoluene 98%
4-Iodo-2-nitrotoluene

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₇H₆INO₂
MW: 263,03 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00051090
CAS-nummer: 41252-97-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR