(R)-N-Boc-piperidine-2-methanol 95%

BLDPBD47299-1GEA 66 EUR
BLDPBD47299-1G BLDPBD47299-5G BLDPBD47299-250MG BLDPBD47299-100MG
(R)-N-Boc-piperidine-2-methanol 95%
(R)-N-Boc-piperidine-2-methanol

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₁H₂₁NO₃
MW: 215,2900 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD04973120
CAS-nummer: 134441-61-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR