(S)-Propane-1,2-diamine dihydrochloride 95%

Leverancier: BLD PHARMATECH GMBH

BLDPBD48416-500GEA 1053 EUR
BLDPBD48416-500G BLDPBD48416-10G BLDPBD48416-0.025G BLDPBD48416-5G BLDPBD48416-100G BLDPBD48416-1G BLDPBD48416-25G
(S)-Propane-1,2-diamine dihydrochloride 95%
(S)-Propane-1,2-diamine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₃H₁₂Cl₂N₂
MW: 147,05 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00050706
CAS-nummer: 19777-66-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR