4-Benzyloxy-2-methylphenylboronic acid 97%

BLDPBD4892-25GEA 344 EUR
BLDPBD4892-25G BLDPBD4892-5G BLDPBD4892-1G BLDPBD4892-250MG BLDPBD4892-10G
4-Benzyloxy-2-methylphenylboronic acid 97%
4-Benzyloxy-2-methylphenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formule: C₁₄H₁₅BO₃
MW: 242,08 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03788404
CAS-nummer: 847560-49-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR