2-Fluoro-p-toluidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD4937-10GEA 13 EUR
BLDPBD4937-10G BLDPBD4937-5G BLDPBD4937-25G BLDPBD4937-1G
2-Fluoro-p-toluidine 98%
2-Fluoro-p-toluidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₇H₈FN
MW: 125,15 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00040975
CAS-nummer: 452-80-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR