6-Iodo-m-toluidine 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD4943-500GEA 1167 EUR
BLDPBD4943-500G BLDPBD4943-250MG BLDPBD4943-5G BLDPBD4943-25G BLDPBD4943-1G BLDPBD4943-10G BLDPBD4943-100G
6-Iodo-m-toluidine 98%
6-Iodo-m-toluidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₇H₈IN
MW: 233,0496 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00833395
CAS-nummer: 13194-69-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR