1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95%

BLDPBD51435-250MGEA 69 EUR
BLDPBD51435-250MG BLDPBD51435-1G
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol 95%
1-(4-(Trifluoromethyl)pyrimidin-2-yl)piperidin-4-ol

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₀H₁₂F₃N₃O
MW: 247,2200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01311906
CAS-nummer: 401930-07-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR