2,6-Dimethoxyphenylboronic acid 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD5278-5GEA 10 EUR
BLDPBD5278-5G BLDPBD5278-100G BLDPBD5278-10G BLDPBD5278-1G BLDPBD5278-500G BLDPBD5278-25G
2,6-Dimethoxyphenylboronic acid 98%
2,6-Dimethoxyphenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₁₁BO₄
MW: 181,98 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01318987
CAS-nummer: 23112-96-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR