1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 95%

BLDPBD5298-10GEA 336 EUR
BLDPBD5298-10G BLDPBD5298-25G BLDPBD5298-1G BLDPBD5298-5G
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate 95%
1-Benzyl-3-methylpiperazine-1,3-dicarboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formule: C₁₄H₁₈N₂O₄
MW: 278,3037 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD03001724
CAS-nummer: 129799-11-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR