1-Amino-2-methylpropan-2-ol 98%

BLDPBD56362-500GEA 1257 EUR
BLDPBD56362-500G BLDPBD56362-1G BLDPBD56362-10G BLDPBD56362-5G BLDPBD56362-100G BLDPBD56362-25G BLDPBD56362-0.025G
1-Amino-2-methylpropan-2-ol 98%
1-Amino-2-methylpropan-2-ol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₄H₁₁NO
MW: 89,1362 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01697283
CAS-nummer: 2854-16-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR