1-Furfurylpyrrole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD58425-25GEA 125 EUR
BLDPBD58425-25G BLDPBD58425-5G BLDPBD58425-1G
1-Furfurylpyrrole 98%
1-Furfurylpyrrole

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formule: C₉H₉NO
MW: 147,18 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00010080
CAS-nummer: 1438-94-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR