1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD58972-5GEA 30 EUR
BLDPBD58972-5G BLDPBD58972-25G BLDPBD58972-10G BLDPBD58972-250MG BLDPBD58972-100G BLDPBD58972-1G
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid 97%
1H-Pyrrolo[2,3-b]pyridine-3-carboxylicacid

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₆N₂O₂
MW: 162,15 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07778360
CAS-nummer: 156270-06-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR