(±)-1-Phenyl-1-propanol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD60122-25GEA 13 EUR
BLDPBD60122-25G BLDPBD60122-1000G BLDPBD60122-5G BLDPBD60122-100G BLDPBD60122-500G
(±)-1-Phenyl-1-propanol 98%
(±)-1-Phenyl-1-propanol

Storage condition: Sealed in dry, Room Temperature

Formule: C₆H₅CH(OH)CH₂CH₃
MW: 136,19 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00004564
CAS-nummer: 93-54-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR