1-tert-Butyl-4-piperidone 98%

BLDPBD60495-250MGEA 22 EUR
BLDPBD60495-250MG BLDPBD60495-5G BLDPBD60495-1G
1-tert-Butyl-4-piperidone 98%
1-tert-Butyl-4-piperidone

Storage condition: Inert atmosphere, Room Temperature

Formule: C₉H₁₇NO
MW: 155,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03411608
CAS-nummer: 1465-76-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR