2-(2,3-Difluorophenyl)ethanol 95%

BLDPBD60920-1GEA 65 EUR
BLDPBD60920-1G BLDPBD60920-5G BLDPBD60920-250MG
2-(2,3-Difluorophenyl)ethanol 95%
2-(2,3-Difluorophenyl)ethanol

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₈F₂O
MW: 158,15 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD01862011
CAS-nummer: 126163-30-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR