2-(Methylthio)thiazole 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD63328-25GEA 46 EUR
BLDPBD63328-25G BLDPBD63328-0.025G BLDPBD63328-100G BLDPBD63328-5G
2-(Methylthio)thiazole 98%
2-(Methylthio)thiazole

Storage condition: Inert atmosphere, Room Temperature

Formule: C₄H₅NS₂
MW: 131,21 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00858981
CAS-nummer: 5053-24-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR