ω,2,4-Trichloroacetophenone 98%

BLDPBD64117-25GEA 7 EUR
BLDPBD64117-25G
ω,2,4-Trichloroacetophenone 98%
ω,2,4-Trichloroacetophenone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₅Cl₃O
MW: 223,49 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00000934
CAS-nummer: 4252-78-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR