2,3,4,5,6-Pentafluorobiphenyl 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD64816-100MGEA 13 EUR
BLDPBD64816-100MG BLDPBD64816-250MG BLDPBD64816-1G BLDPBD64816-5G
2,3,4,5,6-Pentafluorobiphenyl 98%
2,3,4,5,6-Pentafluorobiphenyl

Storage condition: Sealed in dry, Room Temperature

Formule: C₁₂H₅F₅
MW: 244,1601 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00093108
CAS-nummer: 784-14-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR