2,4-Dichloro-5-pyrimidinecarboxylic acid 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD66686-100MGEA 22 EUR
BLDPBD66686-100MG BLDPBD66686-10G BLDPBD66686-1G BLDPBD66686-250MG BLDPBD66686-5G
2,4-Dichloro-5-pyrimidinecarboxylic acid 97%
2,4-Dichloro-5-pyrimidinecarboxylic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formule: C₅H₂Cl₂N₂O₂
MW: 192,9876 g/mol
Opslagtemperatuur: Vriezer
MDL-nummer: MFCD09999133
CAS-nummer: 37131-89-8

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR