2,4-Dichloro-6-nitrophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD66723-5GEA 24 EUR
BLDPBD66723-5G BLDPBD66723-1G BLDPBD66723-25G
2,4-Dichloro-6-nitrophenol 98%
2,4-Dichloro-6-nitrophenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₆H₃Cl₂NO₃
MW: 208 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00007101
CAS-nummer: 609-89-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR