(3-Methoxyphenyl)acetic acid 97%

BLDPBD6697-25GEA 38 EUR
BLDPBD6697-25G BLDPBD6697-5G BLDPBD6697-10G BLDPBD6697-500G BLDPBD6697-100G
(3-Methoxyphenyl)acetic acid 97%
(3-Methoxyphenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formule: (C₆H₄(OCH₃))CH₂COOH
MW: 166,18 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00004334
CAS-nummer: 1798-09-0

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR