2,4-Dimethylimidazool 96%

Leverancier: BLD PHARMATECH GMBH

BLDPBD67113-10GEA 10 EUR
BLDPBD67113-10G BLDPBD67113-5G BLDPBD67113-25G
2,4-Dimethylimidazool 96%
2,4-Dimethylimidazool

Storage condition: Sealed in dry, Room Temperature

Formule: C₅H₈N₂
MW: 96,13 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00022365
CAS-nummer: 930-62-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR