2,5-Dimethoxyphenol 97%

BLDPBD67723-5GEA 365 EUR
BLDPBD67723-5G BLDPBD67723-1G BLDPBD67723-250MG BLDPBD67723-25G
2,5-Dimethoxyphenol 97%
2,5-Dimethoxyphenol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₈H₁₀O₃
MW: 154,1600 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00094237
CAS-nummer: 18113-18-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR