2-Amino-3-methylbenzamide 97%

Leverancier: BLD PHARMATECH GMBH

BLDPBD69929-5GEA 339 EUR
BLDPBD69929-5G BLDPBD69929-250MG BLDPBD69929-1G BLDPBD69929-100MG
2-Amino-3-methylbenzamide 97%
2-Amino-3-methylbenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₈H₁₀N₂O
MW: 150,1800 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD04034517
CAS-nummer: 1885-32-1

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR