2-Amino-5-bromophenol 98%

BLDPBD70371-100GEA 331 EUR
BLDPBD70371-100G BLDPBD70371-500G BLDPBD70371-10G BLDPBD70371-5G BLDPBD70371-25G BLDPBD70371-1G
2-Amino-5-bromophenol 98%
2-Amino-5-bromophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formule: C₆H₆BrNO
MW: 188,0200 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03095028
CAS-nummer: 38191-34-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR