2-Aminobenzo[d]thiazole-6-carboxylic acid 97%

BLDPBD70747-5GEA 38 EUR
BLDPBD70747-5G BLDPBD70747-100G BLDPBD70747-250MG BLDPBD70747-10G BLDPBD70747-25G BLDPBD70747-1G
2-Aminobenzo[d]thiazole-6-carboxylic acid 97%
2-Aminobenzo[d]thiazole-6-carboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formule: C₈H₆N₂O₂S
MW: 194,21 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00054180
CAS-nummer: 93-85-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR