2-Amino-N,N-dimethylacetamide hydrochloride 97%

BLDPBD70872-1GEA 29 EUR
BLDPBD70872-1G BLDPBD70872-10G BLDPBD70872-25G BLDPBD70872-5G BLDPBD70872-250MG BLDPBD70872-100G
2-Amino-N,N-dimethylacetamide hydrochloride 97%
2-Amino-N,N-dimethylacetamide hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formule: C₄H₁₁ClN₂O
MW: 138,5959 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD08060117
CAS-nummer: 72287-77-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR