2-Benzofuranacetic acid 96%

BLDPBD71090-250MGEA 88 EUR
BLDPBD71090-250MG BLDPBD71090-1G BLDPBD71090-5G BLDPBD71090-100MG
2-Benzofuranacetic acid 96%
2-Benzofuranacetic acid

Storage condition: Sealed in dry, 2-8°C

Formule: C₁₀H₈O₃
MW: 176,1687 g/mol
Opslagtemperatuur: Koelkast
MDL-nummer: MFCD00052742
CAS-nummer: 62119-70-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR