4-Methyloctanoic acid 98%

Leverancier: BLD PHARMATECH GMBH

Synoniemen: 4-methylcaprylic acid

BLDPBD7121-1GEA 9 EUR
BLDPBD7121-1G BLDPBD7121-5G BLDPBD7121-25G
4-Methyloctanoic acid 98%
4-Methyloctanoic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₉H₁₈O₂
MW: 158,24 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00051938
CAS-nummer: 54947-74-9

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR