2-Bromo-3,5-dichloroisonicotinic acid 98%

BLDPBD71563-5GEA 93 EUR
BLDPBD71563-5G BLDPBD71563-25G BLDPBD71563-250MG BLDPBD71563-1G
2-Bromo-3,5-dichloroisonicotinic acid 98%
2-Bromo-3,5-dichloroisonicotinic acid

Storage condition: Sealed in dry, Room Temperature

Formule: C₆H₂BrCl₂NO₂
MW: 270,90 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD03411637
CAS-nummer: 343781-56-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR