2-Chlor-4-fluorothiophenol 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD72961-1GEA 22 EUR
BLDPBD72961-1G BLDPBD72961-25G BLDPBD72961-5G
2-Chlor-4-fluorothiophenol 98%
2-Chlor-4-fluorothiophenol

Storage condition: Inert atmosphere, Room Temperature

Formule: C₆H₄ClFS
MW: 162,61 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00052936
CAS-nummer: 175277-99-3

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR