2-Chloro-4-methylbenzaldehyde 98%

BLDPBD73009-1GEA 11 EUR
BLDPBD73009-1G BLDPBD73009-25G BLDPBD73009-5G BLDPBD73009-100G BLDPBD73009-10G
2-Chloro-4-methylbenzaldehyde 98%
2-Chloro-4-methylbenzaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formule: C₈H₇ClO
MW: 154,5936 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD10703515
CAS-nummer: 50817-80-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR