2-Chloro-4-nitrobenzotrifluoride 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD73040-5GEA 23 EUR
BLDPBD73040-5G BLDPBD73040-1G BLDPBD73040-25G BLDPBD73040-100G
2-Chloro-4-nitrobenzotrifluoride 98%
2-Chloro-4-nitrobenzotrifluoride

Storage condition: Sealed in dry, Room Temperature

Formule: C₇H₃ClF₃NO₂
MW: 225,55 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00171532
CAS-nummer: 151504-80-2

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR