2-Chloro-6-methoxybenzonitrile 98%

BLDPBD73396-25GEA 646 EUR
BLDPBD73396-25G BLDPBD73396-250MG BLDPBD73396-10G BLDPBD73396-5G BLDPBD73396-1G BLDPBD73396-0.025G
2-Chloro-6-methoxybenzonitrile 98%
2-Chloro-6-methoxybenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formule: C₈H₆ClNO
MW: 167,5900 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00019743
CAS-nummer: 6575-10-6

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR