2-Ethylimidazool 98%

Leverancier: BLD PHARMATECH GMBH

BLDPBD74704-100GEA 11 EUR
BLDPBD74704-100G BLDPBD74704-10G BLDPBD74704-500G BLDPBD74704-25G BLDPBD74704-1000G
2-Ethylimidazool 98%
2-Ethylimidazool

Storage condition: Sealed in dry, Room Temperature

Formule: C₅H₈N₂
MW: 96,13 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD00005192
CAS-nummer: 1072-62-4

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR